Cleanroom wipes, BlueSorb® 750
Leverandør: BERKSHIRE
BS750.0909.20 BS750.1212.20 BS750.0404.40
BERKBS750.0909.20EA 12850 NOK
BERKBS750.0909.20 BERKBS750.1212.20 BERKBS750.0404.40
Cleanroom wipes, BlueSorb® 750
Papirservietter Renromsservietter Tørkepapir, renrom
These wipes are composed of a hydroentangled nonwoven blend of 55% cellulose and 45% polyester enhanced with a blue colour. This combination of properties provides BlueSorb® 750 with the level of cleanliness, absorbency, abrasion resistance and chemical compatibility required for general wiping and spill control applications.
  • Excellent combination of the synthetic polyester strength and cleanliness with the absorbent characteristics of cellulose
  • Blue colour allows differentiation for guarding against migration into critical areas
  • Smooth, absorbent and