Cleanroom face masks, Choice®
Leverandør: BERKSHIRE
CHFM18040B
BERKCHFM18040BEA 46450 NOK
BERKCHFM18040B
Cleanroom face masks, Choice®
Masker Munnbind, renrom
Berkshire masks offer superior filtration efficiency while providing long wearing comfort for operators.
  • Filter medium: 100% polypropylene
  • Non-woven inner layer and outerfacing
  • Fully enclosed comfortable nosepiece
  • Polyester/spandex earloop provides comfort to wearers
  • Polypropylene spunbound, hydrophobic
People are a significant source of contamination in any critical manufacturing process. Choice® cleanroom face masks are designed to protect the cleanroom or controlled environment from human contamination.
Providing excellent particle and bacterialMore Product Information