Sedimentation cones, Imhoff type
Leverandør: SciLabware
CRF530
BIBBCRF530EA 4900 NOK
BIBBCRF530
Sedimentation cones, Imhoff type
Trakter Immhoff-trakter
Borosilicate glass.
  • Graduated up to 40 ml and with capacity mark at 1000 ml
For determining the amount of sediment in sewage or industrial waste.