Battery Test Chambers with Safety Packages for Aging, Performance or Stress Tests
Leverandør: Binder
9420-0122 9420-0341 9422-0071 9422-0169 9422-0174 9422-0301 9423-0076 9418-0585 9418-0607 9419-0144 9422-0288 9418-0515
BIND9420-0122EA 0 NOK
BIND9420-0122 BIND9420-0341 BIND9422-0071 BIND9422-0169 BIND9422-0174 BIND9422-0301 BIND9423-0076 BIND9418-0585 BIND9418-0607 BIND9419-0144 BIND9422-0288 BIND9418-0515
Battery Test Chambers with Safety Packages for Aging, Performance or Stress Tests
Testkammer Temperaturtestkammer
The test chambers for energy storage systems carrying out aging, stress and performance tests, offer maximum user convenience and are exactly in line with EUCAR Hazard Level 4, 5 or 6. Within a manufacturing process, the test chamber equipped with Package P, P Plus or Package S is perfectly suited for forming the cells.
  • Perfectly suited for temperature tests between 0 and 100 °C
  • Excellent price-performance ratio
  • Compact dimensions compared to other models
  • Small footprint – ideal for small setup areas
  • Choice of 3 different packages for aging, performance