Electrochemistry kit
Leverandør: 3B Scientific
1002711
BINH1002711EA 2925 NOK
BINH1002711
Electrochemistry kit
Lærings materiell Læremateriale for kjemi Electrochemistry Learning Activities
Equipment set inclusive of digital multimeter for measuring electrochemical potentials of various metals in experiments.
Leveringsinformasjon: Kit supplied with a digital multimeter with 2 cables with crocodile clamps, 1× flat trough, 1× copper plate, 1× zinc plate, 1× iron plate, 2× nickel plate, 1× aluminium plate, 2× electrolyte-carbon plates.