Tuning Fork
Leverandør: 3B Scientific
U10115
BINHU10115EA 1075 NOK
BINHU10115
Tuning Fork
Lærings materiell Læremateriale for fysikk Waves and Oscillations Learning Activities
Suitable as a source of intense, high frequency sound, for example, for producing stationary sound waves in Kundt's tube.