Teslameter digital
Leverandør: 3B Scientific
U33110-230
BINHU33110-230EA 11025 NOK
BINHU33110-230
Teslameter digital
Elektromagnetisk feltmåler
This economical digital teslameter will allow students to incorporate quantitative measurements into their magnetism experiments. The unit includes a Hall sensor probe for measuring axial and tangential magnetic fields up to 200 mT. The probe also serves as a ruler as it includes a metric scale.
The teslameter can be calibrated by the user.
In addition to having digital display, the unit outputs a voltage proportional to the magnetic field which can be measured with a data logger, XY-recorder or analogue multimeter.
Bestill nå
SPESIFIKASJONER
Environmentally Preferable
Måleområde 0 - ±20 mT; 0 - ±200 mT
Oppløsning 0,01 mT; 0,1 mT
Probe Hall sensor, 360×15×25 mm, BNC socket
Display LCD 3,5 inch
Utbytte 4 mm safety sockets
B×D×H 205×230×85 mm