Vibration generator
Leverandør: 3B Scientific
U56001
BINHU56001EA 6200 NOK
BINHU56001
Vibration generator
Lærings materiell Læremateriale for fysikk Waves and Oscillations Learning Activities
Vibration generator for exciting oscillating and waves mechanically, e.g. in coil springs, a rubber cord, a wire ring or a Chladni plate. In robust plastic housing including mounting pin with 4 mm socket for attaching accessories (Chladni plates, resonance wire, rubber band etc.). Including holder for stand rod (up to Ø= 8 mm) on the rear side of the apparatus for the demonstration of standing waves in a coil spring. The generator is equipped with overload protection and is driven by a loudspeaker for smooth oscillations and fine travel stepping.
  • Connection: via 4 mm