Tuning fork with plotter pen
Leverandør: 3B Scientific
U8431030
BINHU8431030EA 1375 NOK
BINHU8431030
Tuning fork with plotter pen
Lærings materiell Læremateriale for fysikk Waves and Oscillations Learning Activities
Tuning fork that allows for plotting oscillations on a sheet of paper. The oscillation is triggered by pushing the prongs of the fork together. The oscillation of the tuning fork is highly visible both with the naked eye or with the aid of a stroboscope. Includes a plotter pen with holder and a counterweight.
  • Length: 245 mm
  • Total weight: approx. 170 g