Surface protection papers, LabSorb Ultra
Leverandør: Ahlstrom-Munksjö
144027 144028
BINZ144027EA 2075 NOK
BINZ144027 BINZ144028
Surface protection papers, LabSorb Ultra
Benkepapir Benkepapir
Double-layered LabSorb Ultra papers protect benches and working areas from spilled chemicals, corrosive materials, as well as toxic, infectious or radioactive substances. Spills are quickly absorbed into the surface layer of cellulose material and prevented from penetrating to the surface of the work bench by a thin coating of polyethylene that serves as a carrier for the absorbent layer.
  • Top layer consists of highly absorbent cellulose to quickly capture any spills
  • Bottom layer is an impermeable polyethylene coating that prevents contamination of the