Syringe filters, ReliaMAX™
Leverandør: Ahlstrom-Munksjö
7X7512 7X7531 7X7532 7X7539
BINZ7X7512EA 1700 NOK
BINZ7X7512 BINZ7X7531 BINZ7X7532 BINZ7X7539
Syringe filters, ReliaMAX™
Filter Sprøytefilter
These syringe filters with PP housing contain prefilters specifically developed to increase the efficiency of laboratory microfiltration applications by reducing the number of steps needed to process samples and solutions with either a high particle load or a high viscosity if standard syringe filters are used.
  • Suitable for clarification of solutions, removal of particles and sample preparation
  • High particle holding capacity larger amount of particles can be captured prior to clogging the membrane
  • Fast initial flow – the specific structure of ReliaMAX™