Sample Collection Cards, LeafSaver
Leverandør: Ahlstrom-Munksjö
8.01119.0001BN
BINZ8.01119.0001BNEA 0 NOK
BINZ8.01119.0001BN
Sample Collection Cards, LeafSaver
Diagnostiske systemer Clinical Sample Collection/Preparation Biological Sample Collection/Preparation
This LeafSaver is ideal for simple collection, transportation and ambient-temperature storage of plants, fungi, insects and parasites.
  • DNA-free card for higher accuracy of the results
  • High quality STR profiling and NGS studies
  • Direct-mutiplex PCR and quantitative PCR
  • User-friendly design to facilitate collection, avoid cross-contamination and to protect the biological samples
  • Long-term storage and transportation of samples at ambient temperature.
This card is designed to capture diverse small Eukaryotic organisms, including plants, fungi, insects andMore Product Information