Glutathione Fluorometric Assay Kit

Leverandør: Abcam

K251-100
BIOVK251-100EA 0 NOK
BIOVK251-100
Glutathione Fluorometric Assay Kit
Assay Proteinanalyser

Simple assay to detect total glutathione.


  • Fluorescence (Ex/Em 380/461 nm)
  • Mammalian
  • The Glutathione Detection Kit provides a simple in vitro assay for detecting the GSH changes in apoptosis and other pathological processes


Glutathione is the principal intracellular low-molecular-weight thiol that plays a critical role in the cellular defence against oxidative and nitrosative stress in mammalian cells. Diminished glutathione levels have been observed in the early stages of apoptosis. BioVision’s ApoGSH™ Glutathione Detection Kit provides a simple in vitro assay for detection of total glutathione changes in apoptosis and other samples. The assay utilizes monochlorobimane (MCB), a dye that appears to form an adduct exclusively with glutathione. The unbound MCB is almost nonfluorescent, whereas the dye fluoresces blue (Ex./Em. = 380/461 nm) when bound to glutathione of reduced or oxidized form. The reaction is catalysed by glutathione S-transferase. Thus, the amount of total glutathione can be easily detected using a fluorometer or a 96-well fluorometric plate reader.

Bestill nå


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR