Anti-PHLDA2 Rabbit Polyclonal Antibody (FITC (Fluorescein))

Leverandør: Biorbyt
ORB103663-100
BIRBORB103663-100EA 0 NOK
BIRBORB103663-100
Anti-PHLDA2 Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antistoffer
Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: PHLDA2 (pleckstrin homology-like domain, family A, member 2)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR