Anti-Gentamicin Mouse monoclonal antibody

Leverandør: Biorbyt
ORB10386-100 ORB10386-200
BIRBORB10386-100EA 0 NOK
BIRBORB10386-100 BIRBORB10386-200
Anti-Gentamicin Mouse monoclonal antibody
Antistoffer
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: Gentamicin
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Bestill nå

SPESIFIKASJONER

Environmentally Preferable
Antigen-navn Gentamicin
Antistofftype Primary
Klonalitet Monoclonal
Konjugering Unconjugated
Reaktivitet
Vert Mouse
Isotype IgG
Western blot Yes
ImmunoKjemi Yes
Immunogen Synthetic peptide
Skjema liquid
Rensing purified by affinity chromatography
Konsentrasjon 0,5 mg/ml
Lagringsbuffer 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR