Anti-KLH Rabbit Polyclonal Antibody

Leverandør: Biorbyt
ORB10961-100 ORB10961-200
BIRBORB10961-100EA 0 NOK
BIRBORB10961-100 BIRBORB10961-200
Anti-KLH Rabbit Polyclonal Antibody
Antistoffer
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: KLH (Keyhole Limpet Hemocyanin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

SPESIFIKASJONER

Antigensymbol KLH
Antigen-navn Keyhole Limpet Hemocyanin
Environmentally Preferable
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet
Vert Rabbit
Isotype IgG
Western blot Yes
ImmunoKjemi Yes
Immunogen Synthetic peptide
Skjema liquid
Rensing purified by affinity chromatography
Konsentrasjon 0,5 mg/ml
Lagringsbuffer 10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR