Anti-KLH Mouse Polyclonal Antibody (HRP (Horseradish Peroxidase))

Leverandør: Biorbyt
ORB110276-100
BIRBORB110276-100EA 0 NOK
BIRBORB110276-100
Anti-KLH Mouse Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antistoffer
Type: Primary
Antigen: KLH (Keyhole Limpet Hemocyanin)
Clonality: polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR