Anti-KLH Mouse Polyclonal Antibody (AP (Alkaline Phosphatase))

Leverandør: Biorbyt
ORB113723-100
BIRBORB113723-100EA 0 NOK
BIRBORB113723-100
Anti-KLH Mouse Polyclonal Antibody (AP (Alkaline Phosphatase))
Antistoffer
Type: Primary
Antigen: KLH (Keyhole Limpet Hemocyanin)
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR