Anti-KLH Mouse Polyclonal Antibody (PE (Phycoerythrin)/Cy5.5®)

Leverandør: Biorbyt
ORB130488-100
BIRBORB130488-100EA 0 NOK
BIRBORB130488-100
Anti-KLH Mouse Polyclonal Antibody (PE (Phycoerythrin)/Cy5.5®)
Antistoffer
Type: Primary
Antigen: KLH (Keyhole Limpet Hemocyanin)
Clonality: polyclonal
Clone:
Conjugation: PE (Phycoerythrin)/Cy5.5®
Epitope:
Host: Mouse
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR