Anti-2,4-Dichlorophenoxyacetic acid Rabbit Polyclonal Antibody (FITC (Fluorescein))

Leverandør: Biorbyt
ORB16704-100
BIRBORB16704-100EA 0 NOK
BIRBORB16704-100
Anti-2,4-Dichlorophenoxyacetic acid Rabbit Polyclonal Antibody (FITC (Fluorescein))
Antistoffer
ELISA: 1:500-1000; Western Blot: 1:100-500; Immunochemistry: 1:100-500 (based on 0.5 mg/ml)

Type: Primary
Antigen: 2,4-Dichlorophenoxyacetic acid
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR