Anti-Glyoxalate Reductase Rabbit Polyclonal Antibody

Leverandør: Biorbyt
ORB20804-1
BIRBORB20804-1EA 0 NOK
BIRBORB20804-1
Anti-Glyoxalate Reductase Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: Glyoxalate Reductase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Plant
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR