Anti-Glyoxalate Reductase Rabbit Polyclonal Antibody (Biotin)

Leverandør: Biorbyt
ORB21111-1
BIRBORB21111-1EA 0 NOK
BIRBORB21111-1
Anti-Glyoxalate Reductase Rabbit Polyclonal Antibody (Biotin)
Antistoffer
Type: Primary
Antigen: Glyoxalate Reductase
Clonality: polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Plant
Bestill nå

SPESIFIKASJONER

Antigensymbol GLYR
Environmentally Preferable
Antigen-navn Glyoxalate Reductase
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Biotin
Reaktivitet Plant
Vert Rabbit
Isotype
Immunogen Glyoxylate reductase isolated and purified from spinach. Freunds complete adjuvant is used in the first step of the immunization procedure.
Format immunoconjugate
Konsentrasjon 1 mg/ml, Biotin/IgG protein molar ratio (B/P) approximately 5,4, no foreign proteins added,
Lagringsbuffer phosphate buffered saline
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR