Anti-KLH Rabbit Polyclonal Antibody

Leverandør: Biorbyt
ORB25419-0.2
BIRBORB25419-0.2EA 0 NOK
BIRBORB25419-0.2
Anti-KLH Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: KLH (Keyhole Limpet Hemocyanin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Bestill nå

SPESIFIKASJONER

Antigensymbol KLH
Antigen-navn Keyhole Limpet Hemocyanin
Environmentally Preferable
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet
Vert Rabbit
Isotype
Immunogen KLH (Keyhole Limpet Hemocyanin)
Rensing purified by immunoaffinity chromatography
Konsentrasjon 1,15 mg/ml (OD280)
Lagringsbuffer 0,02M Potassium phosphate, 0,15M Sodium chloride, pH 7,2
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR