Anti-Gentamicin Mouse monoclonal antibody

Leverandør: Biorbyt
ORB26058-1
BIRBORB26058-1EA 0 NOK
BIRBORB26058-1
Anti-Gentamicin Mouse monoclonal antibody
Antistoffer
Type: Primary
Antigen: Gentamicin
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR