Anti-MPI Rabbit Polyclonal Antibody

Leverandør: Biorbyt
ORB41866-100
BIRBORB41866-100EA 0 NOK
BIRBORB41866-100
Anti-MPI Rabbit Polyclonal Antibody
Antistoffer
Type: Primary
Antigen: MPI (mannose phosphate isomerase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Bestill nå

SPESIFIKASJONER

Antigensymbol MPI
Antigen-navn mannose phosphate isomerase
Antistofftype Primary
Klonalitet Polyclonal
Konjugering Unconjugated
Reaktivitet Human
Vert Rabbit
Western blot Yes
Immunogen Recombinant protein fragment contain a sequence corresponding to a region within amino acids 1 and 164 of MPI.
Aminosyretall 1 to 164
Rensing purified by antigen affinity chromatography
Konsentrasjon 1 mg/ml
Lagringsbuffer 0,1M Tris, 0,1M Glycine, 20% Glycerol (pH7).
Lagringstemp Cool
Frakttemp. Ice

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR