Anti-KLH Mouse Monoclonal Antibody

Leverandør: Biorbyt
ORB48769-200
BIRBORB48769-200EA 0 NOK
BIRBORB48769-200
Anti-KLH Mouse Monoclonal Antibody
Antistoffer
Type: Primary
Antigen: KLH (Keyhole Limpet Hemocyanin)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG (Mouse)
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR