Anti-JAK3 Mouse Monoclonal Antibody

Leverandør: Biorbyt
ORB69195-100
BIRBORB69195-100EA 0 NOK
BIRBORB69195-100
Anti-JAK3 Mouse Monoclonal Antibody
Antistoffer
Western Blot: 1:500-1:2000; Immunofluorescence: 1:200-1:1000; Flow Cytometry: 1:200-1:400; ELISA: 1:10000

Type: Primary
Antigen: JAK3 (Janus kinase 3)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR