Anti-AFP Mouse Monoclonal Antibody (CF568) [clone: MBS-12]
Leverandør: Biotium
BNC680521-500 BNC680521-100
BNC680521-500EA 6875 NOK
BNC680521-500 BNC680521-100
Anti-AFP Mouse Monoclonal Antibody (CF568) [clone: MBS-12]
Antistoffer
This antibody recognizes an oncofetal glycoprotein with a single chain of 70 kDa, which is identified as alpha fetoprotein (AFP). This MAb is highly specific to AFP and shows no cross-reaction with other oncofetal antigens or serum albumin. The yolk sac and the liver produce AFP during fetal life. AFP expression in adults is often associated with hepatoma or teratoma. However, hereditary persistence of alpha-fetoprotein may also be found in individuals with no obvious pathology. The protein is thought to be the fetal counterpart of serum albumin, and the AFP and albumin genes are present inMore Product Information