Anti-CDKN1C Mouse Monoclonal Antibody (CF568) [clone: KP10]
Leverandør: Biotium
BNC680879-500 BNC680879-100
BNC680879-500EA 6875 NOK
BNC680879-500 BNC680879-100
Anti-CDKN1C Mouse Monoclonal Antibody (CF568) [clone: KP10]
Antistoffer
Recognizes a protein of 57 kDa, identified as p57Kip2. It shows no cross-reaction with p27Kip1. p57Kip2 is a potent tight-binding inhibitor of several G1 cyclin complexes, and is a negative regulator of cell proliferation. Anti-p57 has been used as an aide in identification of complete hydatidiform mole (CHM) (no nuclear labeling of cytotrophoblasts and stromal cells) from partial hydatidiform mole (PHM) in which both cytotrophoblasts and stromal cells stain. The histological differentiation of complete mole, partial mole, and hydropic spontaneous abortion is problematic. Most completeMore Product Information