Anti-NCAM1 Mouse Monoclonal Antibody (CF488A) [clone: NCAM1/795]

Leverandør: Biotium
BNC880795-500 BNC880795-100
BNC880795-500EA 6875 NOK
BNC880795-500 BNC880795-100
Anti-NCAM1 Mouse Monoclonal Antibody (CF488A) [clone: NCAM1/795]
Antistoffer
This MAb reacts with an extracellular domain (close to transmembrane) of CD56/NCAM. Three isoforms of neural cell adhesion molecule (NCAM) are produced by differential splicing of the RNA transcript from a single gene. The 135 kDa isoform is the basic molecule, which is glycosylated or sialylated to produce the mature species. Anti-CD56 recognizes two proteins of the neural cell adhesion molecule, the basic molecule expressed on most neuroectodermally derived tissues and neoplasms (e.g. retinoblastoma, medulloblastomas, astrocytomas, neuroblastomas, and small cell carcinomas). It is alsoMore Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol NCAM1
Environmentally Preferable
Antigen-navn Neural Cell Adhesion Molecule 1
Antigen synonymer NCAM120|NCAM|NCAM180|Leu-19|MSK39|NKH1|NCAM140|Neural Cell Adhesion Molecule
Antistofftype Primary
Klonalitet Monoclonal
Konjugering CF® 488A
Klon Clone NCAM1/795
Reaktivitet Human
Vert Mouse
Gen-ID 503878
Isotype IgG1 kappa
Western blot Yes
ImmunoKjemi Yes
ImmunoFluorescens Yes
Konsentrasjon 0,1 mg/ml
Lagringsbuffer PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR