Anti-MYC Mouse Monoclonal Antibody (CF594) [clone: 9E10.]

Leverandør: Biotium
BNC940596-500 BNC940596-100
BNC940596-500EA 6875 NOK
BNC940596-500 BNC940596-100
Anti-MYC Mouse Monoclonal Antibody (CF594) [clone: 9E10.]
Antistoffer
This antibody recognizes a transcription factor of 64-67 kDa, identified as c-myc. Its epitope spans between aa 410-419 (EQKLISEEDL) which is a specific portion of an alpha helical region of human c-myc protein. This MAb shows no cross-reaction with v-myc. c-myc is involved in the control of cell proliferation and differentiation and is amplified and/or overexpressed in a variety of tumors. Over-expression of c-myc protein occurs frequently in luminal cells of prostate intraepithelial neoplasia as well as in most primary carcinomas and metastatic disease.

Type: Primary
Antigen: MYC
More Product Information
Bestill nå

SPESIFIKASJONER

Antigensymbol MYC
Antigen-navn c-Myc
Environmentally Preferable
Antistofftype Primary
Klonalitet Monoclonal
Konjugering CF® 594
Klon Clone 9E10.
Reaktivitet Human
Vert Mouse
Gen-ID 202453
Isotype IgG1 kappa
ImmunoKjemi Yes
ImmunoFluorescens Yes
Konsentrasjon 0,1 mg/ml
Lagringsbuffer PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR