Anti-MYC Mouse Monoclonal Antibody (Purified) [clone: 9E10.]
Leverandør: Biotium
BNUB0596-500 BNUB0596-100
BNUB0596-500EA 6275 NOK
BNUB0596-500 BNUB0596-100
Anti-MYC Mouse Monoclonal Antibody (Purified) [clone: 9E10.]
Antistoffer
This antibody recognizes a transcription factor of 64-67 kDa, identified as c-myc. Its epitope spans between aa 410-419 (EQKLISEEDL) which is a specific portion of an alpha helical region of human c-myc protein. This MAb shows no cross-reaction with v-myc. c-myc is involved in the control of cell proliferation and differentiation and is amplified and/or overexpressed in a variety of tumors. Over-expression of c-myc protein occurs frequently in luminal cells of prostate intraepithelial neoplasia as well as in most primary carcinomas and metastatic disease.
Type: Primary
Antigen: MYC