Anti-MUC5AC Mouse Monoclonal Antibody (Purified, BSA-free) [clone: 45M1]
Leverandør: Biotium
BNUM0031-50
BNUM0031-50EA 5125 NOK
BNUM0031-50
Anti-MUC5AC Mouse Monoclonal Antibody (Purified, BSA-free) [clone: 45M1]
Antistoffer
This MAb recognizes the peptide core of gastric mucin M1 (recently identified as Mucin 5AC). Its epitope is located in the C-terminal cysteine rich part of the peptide core of MUC5AC. The epitope is destroyed by beta-mercaptoethanol but not by periodate treatment. This mucin is present in primary ovarian mucinous cancer but usually absent in colorectal adenocarcinoma, thus showing an expression pattern opposite to MUC2. Together with a panel of antibodies, Anti-MUC5AC may be useful for differential identification of primary mucinous ovarian tumors from colon adenocarcinoma metastaticMore Product Information