Anti-CD33 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: C33/68]
Leverandør: Biotium
BNUM0068-50
BNUM0068-50EA 5125 NOK
BNUM0068-50
Anti-CD33 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: C33/68]
Antistoffer
Recognizes a 67 kDa glycoprotein, which is identified as CD33. It is a transmembrane protein of the sialic acid-binding immunoglobulin-like lectin (Siglec) family. It belongs to the immunoreceptor tyrosine-based inhibitory motif (ITIM)-containing molecules able of recruiting protein tyrosine phosphatases SHP-1 and SHP-2 to signal assemblies; these ITIMs are also used for ubiquitin-mediated removal of the receptor from the cell surface. CD33 is expressed on cells of myelomonocytic lineage, binds sialic acid residues in N- and O-glycans on cell surfaces, and is a therapeutic target for acuteMore Product Information