Anti-NF-L Mouse Monoclonal Antibody (Purified, BSA-free) [clone: NM95]
Leverandør: Biotium
BNUM0095-50
BNUM0095-50EA 5125 NOK
BNUM0095-50
Anti-NF-L Mouse Monoclonal Antibody (Purified, BSA-free) [clone: NM95]
Antistoffer
This MAb is an excellent marker for human cells in xenographic model research. It reacts specifically with human cells. This monoclonal antibody is part of a new panel of reagents, which recognizes subcellular organelles or compartments of human cells. These markers may be useful in identification of these organelles in cells, tissues, and biochemical preparations. MAb NM95 recognizes an antigen associated with the nucleoli in human cells. It can be used to stain the nucleoli in cell or tissue preparations and can be used as a marker of the nucleoli in subcellular fractions. It produces aMore Product Information