Anti-EGP40 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: HSA133]

Leverandør: Biotium
BNUM0133-50
BNUM0133-50EA 5125 NOK
BNUM0133-50
Anti-EGP40 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: HSA133]
Antistoffer
MAb HSA133 stains human liver canaliculi and a subset of hepatocellular carcinomas. In frozen sections, it stains liver canaliculi strongly and may be used as a marker of this hepatic substructure. Cell preparations of hepatocellular carcinoma biopsies and cell lines are found to bind this MAb on the cell surface. HSA133 strongly stains liver canaliculi and hepatic carcinoma cells using frozen sections or paraformaldehyde fixed cell preparations.

Type: Primary
Antigen: EGP40
Clonality: Monoclonal
Clone: HSA133
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR