Anti-ITGAX Mouse Monoclonal Antibody (Purified, BSA-free) [clone: HC1/1]

Leverandør: Biotium
BNUM0152-50
BNUM0152-50EA 5125 NOK
BNUM0152-50
Anti-ITGAX Mouse Monoclonal Antibody (Purified, BSA-free) [clone: HC1/1]
Antistoffer
CD11c, also known as Integrin, alpha X (complement component 3 receptor 4 subunit) (ITGAX), is a type I transmembrane protein found at high levels on most human dendritic cells, but also on monocytes, macrophages, neutrophils, and some B cells that induces cellular activation and helps trigger neutrophil respiratory burst; expressed in hairy cell leukemias, acute non-lymphocytic leukemias, and some B-cell chronic lymphocytic leukemias.

Type: Primary
Antigen: ITGAX
Clonality: Monoclonal
Clone: HC1/1
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR