Anti-GFAP Mouse Monoclonal Antibody (Purified, BSA-free) [clone: GA-5]

Leverandør: Biotium
BNUM0167-50
BNUM0167-50EA 5125 NOK
BNUM0167-50
Anti-GFAP Mouse Monoclonal Antibody (Purified, BSA-free) [clone: GA-5]
Antistoffer
This MAb recognizes a protein of ~50 kDa which is identified as Glial Fibrillary Acidic Protein (GFAP). It shows no cross-reaction with other intermediate filament proteins. GFAP is specifically found in astroglia. GFAP is a very popular marker for localizing benign astrocyte and neoplastic cells of glial origin in the central nervous system. Antibody to GFAP is useful in differentiating primary gliomas from metastatic lesions in the brain and for documenting astrocytic differentiation in tumors outside the CNS.

Type: Primary
Antigen: GFAP
Clonality: Monoclonal
Clone: GA-5
Conjugation:More Product Information
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR