Anti-FN1 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: HFN7.1]
Leverandør: Biotium
BNUM0270-50
BNUM0270-50EA 5125 NOK
BNUM0270-50
Anti-FN1 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: HFN7.1]
Antistoffer
Fibronectin is a dimeric glycoprotein of 440 kDa, which is present in cells, extracellular matrix, and blood. It possesses at least four binding sites for collagen, glycosaminoglycans, transglutaminase, and a cell surface receptor. Fibronectin is involved in cell adhesion, tissue organization, and wound healing. This MAb is directed against the peptide core and reacts with both the plasma and cellular forms of fibronectin. It blocks the fibronectin-medicated cell attachment not by disrupting the collagen-fibronectin interaction, but by interfering with the attachment of fibronectin to itsMore Product Information