Anti-CD53 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: 161-2]
Leverandør: Biotium
BNUM0324-50
BNUM0324-50EA 5125 NOK
BNUM0324-50
Anti-CD53 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: 161-2]
Antistoffer
Recognizes a protein of 33-55 kDa, identified as CD53 (Workshop VI; Code N-L033). It is expressed on monocytes and macrophages, dendritic cells, osteoblasts and osteoclasts, and on T and B cells from every stage of differentiation but is absent from platelets, red blood cells. CD53 appears to be the marker with widest reactivity as well as the marker with the strictest specificity to hematopoietic cells. CD53 is a type III membrane with both termini in the cytoplasm and two loops in the extracellular environment. This molecule, in common with other members of tetraspan family, is involved inMore Product Information