Anti-Bovine Serum Albumin Mouse Monoclonal Antibody (Purified, BSA-free) [clone: ALB/398]
Leverandør: Biotium
BNUM0398-50
BNUM0398-50EA 5125 NOK
BNUM0398-50
Anti-Bovine Serum Albumin Mouse Monoclonal Antibody (Purified, BSA-free) [clone: ALB/398]
Antistoffer
Reacts with a protein of ~66 kDa, identified as bovine serum albumin (BSA). It is a high affinity antibody and can be used for detection of traces of BSA. Bovine serum albumin (BSA) is an abundant plasma protein in cows that is important for maintaining osmotic pressure in blood plasma for proper distribution of body fluids between intravascular compartments and body tissues. BSA is a common buffer component for immunoglobulin type assays due to good solubility characteristics for water, Ca2 , Na , K , fatty acids, hormones and bilirubin. BSA makes up about half of the protein in plasma andMore Product Information