Anti-ESR1 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: AER314]

Leverandør: Biotium
BNUM0406-50
BNUM0406-50EA 5125 NOK
BNUM0406-50
Anti-ESR1 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: AER314]
Antistoffer
This MAb is specific to ER alpha and shows minimal cross-reaction with other members of the family. Epitope of this MAb is mapped between aa120-170. ER is an important regulator of growth and differentiation in the mammary gland. Presence of ER in breast tumors indicates an increased likelihood of response to anti-estrogen (e.g. tamoxifen) therapy.

Type: Primary
Antigen: ESR1
Clonality: Monoclonal
Clone: AER314
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Cow, Human, Mouse, Rabbit, Rat
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR