Anti-PODXL Mouse Monoclonal Antibody (Purified, BSA-free) [clone: 3D3]

Leverandør: Biotium
BNUM0427-50
BNUM0427-50EA 5125 NOK
BNUM0427-50
Anti-PODXL Mouse Monoclonal Antibody (Purified, BSA-free) [clone: 3D3]
Antistoffer
Podocalyxin is a member of the CD34 transmembrane sialomucin family. It is over-expressed on the podocyte foot projections and plays essential roles in kidney development and homeostasis, blood filtration and urine formation. It is also expressed on vascular endothelia, hematopoietic progenitors and a subset of neurons. Overexpression of podocalyxin may be linked to more aggressive tumor behavior. Podocalyxin antibody can identify podocytes in the urine (podocyturia) that may indicate glomerular disease, pre-eclampsia, and other kidney pathology.

Type: Primary
Antigen: PODXL
Clonality:More Product Information
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR