Anti-NEFH Mouse Monoclonal Antibody (Purified, BSA-free) [clone: RT-97]
Leverandør: Biotium
BNUM0454-50
BNUM0454-50EA 5125 NOK
BNUM0454-50
Anti-NEFH Mouse Monoclonal Antibody (Purified, BSA-free) [clone: RT-97]
Antistoffer
This MAb reacts with a 200 kDa protein, identified as heavy sub-unit of neurofilaments (NF-H). Neurofilaments make up the main structural elements of axons and dendrites and are found in neurons, peripheral nerves, and sympathetic ganglion cells. Neurofilaments consist of three major subunits with molecular weights of 68 kDa (NF-L), 160 kDa (NF-M) and 200 kDa (NF-H). Anti-neurofilament stains a number of neural, neuroendocrine, and endocrine tumors. Neuromas, ganglioneuromas, gangliogliomas, ganglioneuroblastomas, and neuroblastomas stain positively for anti-neurofilament. Neurofilaments areMore Product Information