Anti-MUC1 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: E29]

Leverandør: Biotium
BNUM0478-50
BNUM0478-50EA 5125 NOK
BNUM0478-50
Anti-MUC1 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: E29]
Antistoffer
In Western blotting, this antibody recognizes proteins in MW range of 265-400 kDa, identified as different glycoforms of EMA. EMA may provide a protective layer on epithelial cells against bacterial and enzyme attack. In immunohistochemical assays, it superbly stains routine formalin/paraffin carcinomas. Antibody to EMA is useful as a pan-epithelial marker for detecting early metastatic loci of carcinoma in bone marrow or liver.

Type: Primary
Antigen: MUC1
Clonality: Monoclonal
Clone: E29
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR