Anti-FUT4 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: FR4A5]

Leverandør: Biotium
BNUM0504-50
BNUM0504-50EA 5125 NOK
BNUM0504-50
Anti-FUT4 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: FR4A5]
Antistoffer
CD15 plays a role in mediating phagocytosis, bactericidal activity, and chemotaxis. It is present on  more than 95% of granulocytes including neutrophils and eosinophils and to a lesser degree on monocytes. In addition, CD15 is expressed in Reed-Sternberg cells and some epithelial cells. CD15 antibody is very useful in the identification of Hodgkin’s disease. CD15 is occasionally expressed in large cell lymphomas of both B and T phenotypes which otherwise have a quite distinct histological appearance.

Type: Primary
Antigen: FUT4
Clonality: Monoclonal
Clone: FR4A5
Conjugation: Purified,More Product Information
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR