Anti-CSF2 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: BVD2-21C11]

Leverandør: Biotium
BNUM0662-50
BNUM0662-50EA 5125 NOK
BNUM0662-50
Anti-CSF2 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: BVD2-21C11]
Antistoffer
Granulocyte/macrophage - Colony-stimulating factor (GM-CSF) is a hematopoietic factor that is produced by activated T-cells, B-cells, mast cells, macrophages, fibroblasts, and endothelial cells. In addition to supporting colony formation of granulocyte/macrophage progenitors, GM-CSF is a growth factor for erythroid, megakaryocyte, and eosinophil progenitors.

Type: Primary
Antigen: CSF2
Clonality: Monoclonal
Clone: Clone BVD2-21C11
Conjugation: Unconjugated, BSA-and azide-free
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Cynomolgus monkey, Human, Rhesus monkey
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR