Anti-Il6 (Mouse) Rat Monoclonal Antibody (Purified, BSA-free) [clone: MP5-20F3]

Leverandør: Biotium
BNUM0663-50
BNUM0663-50EA 5125 NOK
BNUM0663-50
Anti-Il6 (Mouse) Rat Monoclonal Antibody (Purified, BSA-free) [clone: MP5-20F3]
Antistoffer
IL-6 is a potent lymphoid cell growth factor that stimulates the growth and survivability of certain B-cells and T-cells. IL-6 plays a role in host defense, acute phase reactions, immune responses, and hematopoiesis. IL-6 is expressed by T-cells, B-cells, monocytes, fibroblasts, hepatocytes, endothelial cells and keratinocytes.

Type: Primary
Antigen: Il6 (Mouse)
Clonality: Monoclonal
Clone: Clone MP5-20F3
Conjugation: Unconjugated, BSA-and azide-free
Epitope:
Host: Rat
Isotype: IgG1 kappa
Reactivity: Mouse
Bestill nå

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR