Anti-MLANA Mouse Monoclonal Antibody (Purified, BSA-free) [clone: M2-7C10 M2-9E3]
Leverandør: Biotium
BNUM0705-50
BNUM0705-50EA 5125 NOK
BNUM0705-50
Anti-MLANA Mouse Monoclonal Antibody (Purified, BSA-free) [clone: M2-7C10 M2-9E3]
Antistoffer
This antibody recognizes a protein doublet of 20-22 kDa, identified as MART-1 (Melanoma Antigen Recognized by T cells 1) or Melan-A. MART-1 is a newly identified melanocyte differentiation antigen recognized by autologous cytotoxic T lymphocytes. Seven other melanoma associated antigens recognized by autologous cytotoxic T cells include MAGE-1, MAGE-3, tyrosinase, gp100, gp75, BAGE-1, and GAGE-1. Subcellular fractionation shows that MART-1 is present in melanosomes and endoplasmic reticulum. This MAb labels melanomas and other tumors showing melanocytic differentiation. It is also a usefulMore Product Information