Anti-KRT7 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: KRT7/760]

Leverandør: Biotium
BNUM0760-50
BNUM0760-50EA 5125 NOK
BNUM0760-50
Anti-KRT7 Mouse Monoclonal Antibody (Purified, BSA-free) [clone: KRT7/760]
Antistoffer
This antibody recognizes an intermediate filament protein (IFP) of 55 kDa, which is identified as cytokeratin 7. This MAb is highly specific to cytokeratin 7 and shows no cross-reaction with other IFPs. Cytokeratin 7 is a basic cytokeratin, which is found in most glandular and transitional epithelia but not in the stratified squamous epithelia. Keratin 7 is expressed in the epithelial cells of ovary, lung, and breast but not of colon, prostate, or gastrointestinal tract. This MAb is highly useful in distinguishing ovarian carcinomas (keratin 7 ) from colon carcinomas (keratin 7-).

Bestill nå

SPESIFIKASJONER

Antigensymbol KRT7
Environmentally Preferable
Antigen-navn Cytokeratin 7
Antigen synonymer KRT7|Krt2-7|Sarcolectin|SCL|K2C7|Keratin 55K Type II Cytoskeletal|CK-7|Type-II Keratin Kb7|Type II Mesothelial Keratin K7|Keratin Simple Epithelial Type 1 K7|Keratin Type II Cytoskeletal 7
Antistofftype Primary
Klonalitet Monoclonal
Konjugering Unconjugated
Klon Clone KRT7/760
Reaktivitet Human
Vert Mouse
Gen-ID 411501
Isotype IgG1 kappa
ImmunoKjemi Yes
ImmunoFluorescens Yes
Rensing BSA-free
Konsentrasjon 1 mg/mL
Lagringsbuffer PBS/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Les mer About VWR